Merck
Tetartohedral twinning could happen to you too.
Acta crystallographica. Section D, Biological crystallography (2012-04-17)
Pietro Roversi, Eric Blanc, Steven Johnson, Susan Mary Lea
ABSTRACT
Tetartohedral crystal twinning is discussed as a particular case of (pseudo)merohedral twinning when the number of twinned domains is four. Tetartohedrally twinned crystals often possess pseudosymmetry, with the rotational part of the pseudosymmetry operators coinciding with the twinning operators. Tetartohedrally twinned structures from the literature are reviewed and the recent structure determination of tetartohedrally twinned triclinic crystals of human complement factor I is discussed.
Sigma-Aldrich
Complement factor I from human plasma, >90% (SDS-PAGE)