Merck
The biology of hepatocyte growth factor/scatter factor.
BioEssays : news and reviews in molecular, cellular and developmental biology (1992-09-01)
R A Furlong
ABSTRACT
Hepatocyte growth factor, a potent mitogen for epithelial and other cell types, and scatter factor, a stimulant of epithelial cell motility are identical. In addition to these mitogenic and motogenic functions, the factor has been shown to be an epithelial morphogen and also has antiproliferative effects in some cancer cell lines. The membrane receptor for hepatocyte growth factor/scatter factor has been identified as the c-met proto-oncogene product.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture