Merck
  • S14G-Humanin ameliorates ovalbumin-induced airway inflammation in asthma mediated by inhibition of toll-like receptor 4 (TLR4) expression and the nuclear factor κ-B (NF-κB)/early growth response protein-1 (Egr-1) pathway.
S14G-Humanin ameliorates ovalbumin-induced airway inflammation in asthma mediated by inhibition of toll-like receptor 4 (TLR4) expression and the nuclear factor κ-B (NF-κB)/early growth response protein-1 (Egr-1) pathway.
Aging (2023-07-15)
Bo Su, Ran Li, Fuxing Song, Min Liu, Xianjun Sun
ABSTRACT
Asthma is a chronic inflammatory disease with a high morbidity rate in children and significantly impacts their healthy growth. It is reported that Th2 cell-mediated airway inflammation and activated oxidative stress are involved in the pathogenesis of asthma. S14G-humanin (HNG) is a derivative of Humanin with higher activity. The present study proposes to explore the potential treating property of HNG on asthma. An asthma model was constructed in mice using ovalbumin (OVA), the mice were treated with 2.5 mg/kg and 5 mg/kg HNG for 16 days. Dramatically increased lung weight index, elevated number of monocytes, eosinophils, and neutrophils, promoted production of Th2 cytokines including interleukin-4 (IL-4), interleukin-5 (IL-5), and interleukin-13 (IL-13), and severe histological pathology were observed in OVA-challenged mice, all of which were extremely alleviated by 2.5 mg/kg and 5 mg/kg HNG. Furthermore, the increased malondialdehyde (MDA) level and declined superoxide dismutase (SOD) activity in OVA-challenged mice were abolished by 2.5 mg/kg and 5 mg/kg HNG. Lastly, the upregulated TLR4, p-NF-κB p65, and early growth response 1 (Egr-1) in lung tissues of OVA-challenged mice were pronouncedly downregulated by 2.5 mg/kg and 5 mg/kg HNG. Collectively, our data suggested that HNG ameliorated airway inflammation in asthma partially due to NF-κB and Egr-1-mediated responses.
Sigma-Aldrich
[Gly14]-Humanin G human, ≥95% (HPLC), powder