Merck
  • Development of a three-dimensional blood-brain barrier network with opening capillary structures for drug transport screening assays.
Development of a three-dimensional blood-brain barrier network with opening capillary structures for drug transport screening assays.
Materials today. Bio (2022-06-28)
Marie Piantino, Dong-Hee Kang, Tomomi Furihata, Noriyuki Nakatani, Kimiko Kitamura, Yukari Shigemoto-Mogami, Kaoru Sato, Michiya Matsusaki
ABSTRACT
The blood-brain barrier (BBB), a selective barrier regulating the active and passive transport of solutes in the extracellular fluid of the central nervous system, prevents the delivery of therapeutics for brain disorders. The BBB is composed of brain microvascular endothelial cells (BMEC), pericytes and astrocytes. Current in vitro BBB models cannot reproduce the human structural complexity of the brain microvasculature, and thus their functions are not enough for drug assessments. In this study, we developed a 3D self-assembled microvascular network formed by BMEC covered by pericytes and astrocyte end feet. It exhibited perfusable microvasculature due to the presence of capillary opening ends on the bottom of the hydrogel. It also demonstrated size-selective permeation of different molecular weights of fluorescent-labeled dextran, as similarly reported for in vivo rodent brain, suggesting the same permeability with actual in vivo brain. The activity of P-glycoprotein efflux pump was confirmed using the substrate Rhodamine 123. Finally, the functionality of the receptor-mediated transcytosis, one of the main routes for drug delivery of large molecules into the brain, could be validated using transferrin receptor (TfR) with confocal imaging, competition assays and permeability assays. Efficient permeability coefficient (Pe) value of transportable anti-TfR antibody (MEM-189) was seven-fold higher than those of isotype antibody (IgG1) and low transportable anti-TfR antibody (13E4), suggesting a higher TfR transport function than previous reports. The BBB model with capillary openings could thus be a valuable tool for the screening of therapeutics that can be transported across the BBB, including those using TfR-mediated transport.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture