Merck
  • Effect of simvastatin on cetuximab resistance in human colorectal cancer with KRAS mutations.
Effect of simvastatin on cetuximab resistance in human colorectal cancer with KRAS mutations.
Journal of the National Cancer Institute (2011-03-15)
Jeeyun Lee, Inkyoung Lee, Boram Han, Joon Oh Park, Jiryeon Jang, Chaehwa Park, Won Ki Kang
ABSTRACT
Metastatic colorectal cancer (CRC) patients with v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog (KRAS) mutations are resistant to treatment with cetuximab, a monoclonal antibody that targets the epidermal growth factor receptor. Statins have reported antitumor activity, but it is unknown whether simvastatin can reverse cetuximab resistance in KRAS mutant CRC. Human CRC cell lines with KRAS mutations (LS153, LS174T, DLD1, LoVo, SW403, SW480, SNU175, and LS1034) or with v-raf murine sarcoma viral oncogene homolog B1 (BRAF) mutations (DiFi, SW48, HT29, and RKO) were used to test the effect of cetuximab, simvastatin, and cetuximab plus simvastatin on cell proliferation and apoptosis in vitro. Because BRAF(V600E) mutant may be responsible for cetuximab resistance in KRAS wild-type cells, we measured the growth of xenograft tumors originating from KRAS mutant and BRAF mutant cells in mice treated with cetuximab alone or plus simvastatin (n = 5 mice per treatment group). We used immunoblot assays to study RAS-regulated activation of BRAF protein after simvastatin treatment. All statistical tests were two-sided. Addition of simvastatin (0.2 μM) to cetuximab (0.03-1.0 μM) reduced cell proliferation of KRAS mutant (P < .001) but not of BRAF mutant CRC cells in vitro. Treatment of KRAS mutant cells with simvastatin reduced BRAF activity and induced apoptosis. Treatment with cetuximab and simvastatin reduced the growth of xenograft tumors originating from KRAS mutant cells compared with cetuximab alone (eg, for tumors originating from DLD1 cells, cetuximab vs cetuximab + simvastatin, mean tumor volume = 49.4 vs 20.2 cm(3), mean difference = 29.2 cm(3), 95% confidence interval = 19.7 to 38.5, P < .001); treatment with cetuximab alone or in combination with simvastatin had no effect on the growth of BRAF mutant tumors. Simvastatin may overcome cetuximab resistance in colon cancer cells with KRAS mutations by modulating BRAF activity and inducing apoptosis.
Sigma-Aldrich
MEK1 Protein, inactive, 50 g, Unactive, N-terminal GST & C-terminal His6-tagged, recombinant full-length human MEK1, for use in Kinase Assays.