- Immune parameters in two different laying hen strains during five production periods.
Immune parameters in two different laying hen strains during five production periods.
During life, the number and function of immune cells change with potential consequences for immunocompetence of an organism. In laying hens, studies have primarily focused on early development of immune competence and only few have investigated systemic and lymphatic distribution of leukocyte subsets during adolescence and the egg-laying period. The present study determined the number of various leukocyte types in blood, spleen, and cecal tonsils of 10 Lohmann Brown-Classic and 10 Lohmann LSL-Classic hens per wk of life 9/10, 15/16, 23/24, 29/30, and 59/60, encompassing important production as well as developmental stages, by flow cytometry. Although immune traits differed between the 2 hen strains, identical patterns of age-related immunological changes were found. The numbers of all investigated lymphocyte types in the spleen as well as the numbers of blood γδ T cells increased from wk 9/10 to 15/16. This suggests an ongoing release of lymphocytes from primary lymphoid tissues and an influx of blood lymphocytes into the spleen due to novel pathogen encounters during adolescence. A strong decrease in the number of CTL and γδ T cells and an increase in innate immune cells within blood and spleen were found between wk of life 15/16 and 23/24, covering the transition phase to egg-laying activity. Numbers of peripheral and splenic lymphocytes remained low during the egg-laying period or even further decreased, for example blood CD4+ T cells and splenic γδ T cells. Functional assessments showed that in vitro IFN-γ production of mitogen-stimulated splenocytes was lower in wk 60. Taken together, egg-laying activity seems to alter the immune system toward a more pronounced humoral and innate immune response, with probable consequences for the immunocompetence and thus for productivity, health and welfare of the hens.