Merck
  • [Kinetic study on inhibition effects of dansyl-L-phenylalanine and L-phenylalanine on calf intestinal alkaline phosphatase].
[Kinetic study on inhibition effects of dansyl-L-phenylalanine and L-phenylalanine on calf intestinal alkaline phosphatase].
Guang pu xue yu guang pu fen xi = Guang pu (2009-12-30)
Li-Na Li, Yu-Qing Wu, René Buchet
ABSTRACT
To evaluate the inhibition effect of dansyl-L-phenylalanine on calf intestinal alkaline phosphatase (CIAP), UV-Vis spectrophotometric method was employed. It was found that dansyl-L-phenylalanine can selectively inhibit CIAP. The kinetic inhibition processes of dansyl-L-phenylalanine and L-phenylalanine were comparatively studied. The authors' finding elucidates that at the optimized alkaline pH of alkaline phosphatase (pH 10.4) and 37 degrees C, dansyl-L-phenylalanine can inhibit alkaline phosphatase activity of CIAP efficiently and specifically, similar as L-phenylalanine. Both inhibition types were uncompetitive inhibition resulting from the double reciprocal curve fitting of upsilon versus substrate concentrations, and the inhibition constants Ki of both inhibitors were determined to be 2.3 and 1.1 mmol L(-1) respectively, both of which were at millimolar level. The investigation of the inhibition effect of dansyl modified L-phenylalanine on calf intestinal alkaline phosphatase not only helped get insight into the detailed inhibition mechanism of L-phenylalanine on tissue specific alkaline phosphatase, such as in the case of intestinal alkaline phosphatase, but also provided the possibility to employ fluorescence spectroscopy by labeling the specific inhibitors of alkaline phosphatase with chromophoric groups.
Sigma-Aldrich
Phosphatase, Alkaline−Agarose from calf intestine, ammonium sulfate suspension