Merck
  • Influence of aqueous film forming foams on the solubility and mobilization of non-aqueous phase liquid contaminants in quartz sands.
Influence of aqueous film forming foams on the solubility and mobilization of non-aqueous phase liquid contaminants in quartz sands.
Water research (2021-03-08)
Shuchi Liao, Zachary Saleeba, J Daniel Bryant, Linda M Abriola, Kurt D Pennell
ABSTRACT
At sites where aqueous film forming foams (AFFFs) are used for fire suppression or training activities, interactions between dissolved foam constituents and organic liquids could alter contaminant migration in the subsurface. In this study, batch reactor and column experiments were conducted to investigate the potential for AFFF solutions to enhance the solubility and mobility of three representative non-aqueous phase liquid (NAPLs), JP-4 jet fuel, trichloroethene (TCE), and tetrachloroethene (PCE). For AFFF concentrations up to 5% wt. (50,000 mg/L), aqueous solubilities of TCE and PCE increased by less than 50%, indicating the absence of micellar solubilization. However, NAPL-water interfacial tensions were reduced to less than 1.5 mN/m and resulted in accumulation of up to 2.25 mg/m2 of AFFF at the NAPL-water interface. To assess the potential for AFFF to mobilize residual (entrapped) NAPL at a field application rate of 3% wt. (30,000 mg/L), columns were packed with two size fractions of Ottawa sands (20-30 mesh and 60-80 mesh) that yielded residual NAPL saturations ranging from 11.7 to 17.6%. Following injection of 3 pore volumes of the 3% wt. AFFF solution, partial mobilization of residual NAPL was observed for PCE, TCE, and JP-4, with saturation reductions of 0.7 to 2% in 20-30 mesh and 0.3% to 1.3% in 60-80 mesh Ottawa sand. The columns were then flushed with an ultralow-IFT surfactant solution consisting of 4% wt. 1:1 Aerosol AY/OT, which resulted in nearly complete mobilization of the remaining residual NAPL. When NAPL desaturation curves were expressed in terms of the total trapping number (NT), the threshold value of NT required for NAPL mobilization by 3% wt. AFFF was approximately 2 × 10-5, consistent with previous studies. These findings demonstrate that AFFF solutions can lead to partial mobilization of residual NAPL, and that the total trapping number concept can be used to predict such behavior a priori. In addition, the observed IFT reductions are sufficient to alter NAPL accumulation and redistribution behavior in the subsurface through enhanced spreading above the water table (e.g., JP-4) or penetration into lower-permeability media (e.g., TCE, PCE).