Merck
  • Genetic Variants Implicate Dual Oxidase-2 in Familial and Sporadic Nonmedullary Thyroid Cancer.
Genetic Variants Implicate Dual Oxidase-2 in Familial and Sporadic Nonmedullary Thyroid Cancer.
Cancer research (2019-09-11)
Darrin V Bann, Qunyan Jin, Kathryn E Sheldon, Kenneth R Houser, Lan Nguyen, Joshua I Warrick, Maria J Baker, James R Broach, Glenn S Gerhard, David Goldenberg
ABSTRACT
Highly penetrant hereditary thyroid cancer manifests as familial nonmedullary thyroid cancer (FNMTC), whereas low-penetrance hereditary thyroid cancer manifests as sporadic disease and is associated with common polymorphisms, including rs965513[A]. Whole-exome sequencing of an FNMTC kindred identified a novel Y1203H germline dual oxidase-2 (DUOX2) mutation. DUOX2Y1203H is enzymatically active, with increased production of reactive oxygen species. Furthermore, patients with sporadic thyroid cancer homozygous for rs965513[A] demonstrated higher DUOX2 expression than heterozygous rs965513[A/G] or homozygous rs965513[A]-negative patients. These data suggest that dysregulated hydrogen peroxide metabolism is a common mechanism by which high- and low-penetrance genetic factors increase thyroid cancer risk. SIGNIFICANCE: This study provides novel insights into the genetic and molecular mechanisms underlying familial and sporadic thyroid cancers.
Sigma-Aldrich
Anti-Dual oxidase 2, clone Duox S-12 Antibody, clone Duox S-12, from mouse