Merck
  • Loss of FOXC1 contributes to the corneal epithelial fate switch and pathogenesis.
Loss of FOXC1 contributes to the corneal epithelial fate switch and pathogenesis.
Signal transduction and targeted therapy (2021-01-09)
Mingsen Li, Liqiong Zhu, Jiafeng Liu, Huaxing Huang, Huizhen Guo, Li Wang, Lingyu Li, Sijie Gu, Jieying Tan, Jing Zhong, Bowen Wang, Zhen Mao, Yong Fan, Chunqiao Liu, Jin Yuan, Hong Ouyang
ABSTRACT
Forkhead box C1 (FOXC1) is required for neural crest and ocular development, and mutations in FOXC1 lead to inherited Axenfeld-Rieger syndrome. Here, we find that FOXC1 and paired box 6 (PAX6) are co-expressed in the human limbus and central corneal epithelium. Deficiency of FOXC1 and alternation in epithelial features occur in patients with corneal ulcers. FOXC1 governs the fate of the corneal epithelium by directly binding to lineage-specific open promoters or enhancers marked by H3K4me2. FOXC1 depletion not only activates the keratinization pathway and reprograms corneal epithelial cells into skin-like epithelial cells, but also disrupts the collagen metabolic process and interferon signaling pathways. Loss of interferon regulatory factor 1 and PAX6 induced by FOXC1 dysfunction is linked to the corneal ulcer. Collectively, our results reveal a FOXC1-mediated regulatory network responsible for corneal epithelial homeostasis and provide a potential therapeutic target for corneal ulcer.
Sigma-Aldrich
Anti-ZNF750 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2