Merck
  • Proteomics and metabolomics analysis of tomato fruit at different maturity stages and under salt treatment.
Proteomics and metabolomics analysis of tomato fruit at different maturity stages and under salt treatment.
Food chemistry (2019-12-31)
Huimeng Tang, Xu Zhang, Biao Gong, Yanyan Yan, Qinghua Shi
ABSTRACT
Proteomics and metabolomics were used to study the changes in proteins and metabolites in tomato fruits at different ripening stages and the effect of salt treatment on fruit quality. The results showed 2607 and 153 differentially expressed proteins in ripe fruits compared with mature green fruits and in NaCl-treated ripe fruits compared with control ripe fruits, respectively. KEGG analysis indicated that these proteins were mainly involved in photosynthesis, pentose and glucuronate interconversions in different ripening stages of fruits, and salt-induced proteins were involved in flavonoid biosynthesis and linoleic acid metabolism. A series of metabolites, including carbohydrates and amino acids showed significantly different accumulations between ripe and mature green fruits and between salt-treated and control fruits. Combined analysis explored glycine, L-alanine, D-xylose and sucrose and some proteins involved in multiple metabolic pathways under salt conditions. Their interactions might affect fruit development and fruit quality under salt treatment.
Product Description
Millipore
Protease Inhibitor Cocktail Set VI, The Protease Inhibitor Cocktail Set VI controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.