Merck
  • Dps protein is related to resistance of Mycobacterium abscessus subsp. massiliense against stressful conditions.
Dps protein is related to resistance of Mycobacterium abscessus subsp. massiliense against stressful conditions.
Applied microbiology and biotechnology (2020-04-08)
Nayra Rodrigues de Alcântara, Fábio Muniz de Oliveira, Wanius Garcia, Otavio Augusto Leitão Dos Santos, Ana Paula Junqueira-Kipnis, André Kipnis
ABSTRACT
Mycobacterium abscessus subsp. massiliense (Mycma) belongs to the Mycobacterium abscessus complex and is a rapidly growing non-tuberculous mycobacterium. The chronic pulmonary, skin, and soft tissue infections that it causes may be difficult to treat due to its intrinsic resistance to the commonly used antimicrobial drugs, making it a serious world public health problem. Iron is an essential nutrient for the growth of microorganisms; nonetheless, it can be toxic when in excess. Thus, bacteria require an iron homeostasis mechanism to succeed in different environments. DNA-binding proteins from starved cells (Dps) are miniferritins with the property to act as additional iron storage proteins but also can bind to DNA, protecting it against hydroxyl radical. Annotation of the Mycma genome revealed the gene mycma_03135 with 79% sequential identity when compared to MSMEG_3242 gene from M. smegmatis mc2 155, which codifies for a known Dps. Recombinant Dps from M. abscessus (rMaDps) was produced in Escherichia coli, purified in soluble form and shown to form high mass oligomers in solution with ferroxidase activity, DNA binding, and protection against damage. The expression of the mycma_03135 gene was induced during Mycma growth in the presence of hydrogen peroxide (H2O2). Additionally, the expression of rMaDps by E. coli conferred greater resistance to H2O2. Thus, this study is the first to identify and characterize a Dps from M. abscessus. KEY POINTS: Mycobacterium abscessus subsp. massiliense express a miniferritin protein (Dps). Mycma Dps binds to DNA and protects against oxidative stress.
Sigma-Aldrich
(±)-Geosmin, ≥97% (GC)