Merck
  • Antimicrobial activity of mesenchymal stem cells against Staphylococcus aureus.
Antimicrobial activity of mesenchymal stem cells against Staphylococcus aureus.
Stem cell research & therapy (2020-07-19)
Haruyo Yagi, Antonia F Chen, David Hirsch, Adam C Rothenberg, Jian Tan, Peter G Alexander, Rocky S Tuan
ABSTRACT
There have been limited advances in the treatment of bone and joint infections, which currently involves a combination of surgery and antibiotic administration. There is a timely need in orthopedics to develop more effective and less invasive forms of antimicrobial prophylaxis and treatment. The antibacterial effect of adult tissue-derived mesenchymal stem cells (MSCs) has recently been investigated against Escherichia coli and Staphylococcus aureus. The main mechanism of action is postulated to be via MSC production of the cationic antimicrobial peptide, LL-37. This study examines the antimicrobial activity of adipose-derived human MSCs (ASCs) on S. aureus, specifically examining the role of LL-37 and regulation of its expression. Bacteria colony-forming unit (CFU) assay was used to assess antimicrobial activity. Our results showed that the ASC-conditioned medium significantly inhibited the growth of S. aureus under standard culture conditions with or without the continued presence of ASCs. Also, the treatment of ASCs with 1,25-dihydroxy vitamin D3 elevated LL-37 expression and enhanced their antimicrobial activity. In support, treatment with the vitamin D receptor inhibitor, GW0742, blocked the antimicrobial activity of ASCs. Our findings clearly demonstrate the antimicrobial activity of adult ASCs against S. aureus and implicate a key regulatory role for vitamin D. Further testing in in vivo models is being pursued to assess the potential application of ASCs as a biocompatible, adjunct treatment for musculoskeletal infections.
Sigma-Aldrich
GW0742, ≥98% (HPLC)