Merck
  • Optimization of the separation lactic acid enantiomers in body fluids by capillary electrophoresis.
Optimization of the separation lactic acid enantiomers in body fluids by capillary electrophoresis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2002-02-05)
L Saavedra, C Barbas
ABSTRACT
The optimization of the separation conditions of the two optical isomers of lactic acid by a factorial design is reported. Initially, different chiral selectors were systematically investigated and then a experimental design with three quantitative factors (cyclodextrin concentration and background buffer pH and concentration) were evaluated. Optimal conditions for obtaining a resolution higher than 1.5 were: phosphate buffer 200 mM at pH=6.0 with 413 mM 2-hydroxypropyl-beta-cyclodextrin added (HP-beta-CD), 20 degrees C, -20 kV of applied potential and polyacrylamide-coated capillary. The method was validated for the measurement in plasma and it was applied to the identification of both isomers in body fluids such as urine, amniotic fluid and cerebrospinal fluid. Samples were centrifuged and diluted (1:4) prior to the analysis.
Sigma-Aldrich
(2-Hydroxyethyl)-β-cyclodextrin, extent of labeling: ~0.7 mol per mol cellulose