Merck
  • Synthesis and central nervous system depressant effects of N3-substituted 2',3'-O-isopropylideneuridines.
Synthesis and central nervous system depressant effects of N3-substituted 2',3'-O-isopropylideneuridines.
Chemical & pharmaceutical bulletin (2000-04-05)
C S Yao, T Kimura, K Watanabe, S Kondo, I K Ho, I Yamamoto
ABSTRACT
N3-Substituted derivatives of 2',3'-O-isopropylideneuridine (1) were synthesized and their pharmacological effects on the central nervous system (CNS) examined using mice. Methyl (2), ethyl (3), propyl (4), butyl (5), allyl (6), benzyl (7), o-, m-, p-xylyls (8, 9, 10), and alpha-phenylethyl (11) derivatives of 1 were administered to mice by intracerebroventricular (i.c.v.) injection for evaluating hypnotic activity, pentobarbital-induced sleep prolongation, and spontaneous activity as indices. Only 3 possessed hypnotic activity by i.c.v. injection at the dose of 2.0 mumol/mouse. Compounds 3, 4, and 10 significantly showed synergism with a barbiturate, indicating that the derivatives have some CNS depressant effects. Moreover, 3 and 4 caused decrease in the spontaneous activity of mice, even at low doses. The present study indicated that substitution by ethyl, propyl, and p-xylyl groups at the N3-position of 2',3'-O-isopropylideneuridine imparted the CNS depressant effects.
Sigma-Aldrich
2′,3′-O-Isopropylideneuridine, ≥99% (HPLC)