Merck
  • HMR 1098 is not an SUR isotype specific inhibitor of heterologous or sarcolemmal K ATP channels.
HMR 1098 is not an SUR isotype specific inhibitor of heterologous or sarcolemmal K ATP channels.
Journal of molecular and cellular cardiology (2010-12-28)
Hai Xia Zhang, Alejandro Akrouh, Harley T Kurata, Maria Sara Remedi, Jennifer S Lawton, Colin G Nichols
ABSTRACT
Murine ventricular and atrial ATP-sensitive potassium (K(ATP)) channels contain different sulfonylurea receptors (ventricular K(ATP) channels are Kir6.2/SUR2A complexes, while atrial K(ATP) channels are Kir6.2/SUR1 complexes). HMR 1098, the sodium salt of HMR 1883 {1-[[5-[2-(5-chloro-o-anisamido)ethyl]-2-methoxyphenyl]sulfonyl]-3-methylthiourea}, has been considered as a selective sarcolemmal (i.e. SUR2A-dependent) K(ATP) channel inhibitor. However, it is not clear whether HMR 1098 would preferentially inhibit ventricular K(ATP) channels over atrial K(ATP) channels. To test this, we used whole-cell patch clamp techniques on mouse atrial and ventricular myocytes as well as (86)Rb(+) efflux assays and excised inside-out patch clamp techniques on Kir6.2/SUR1 and Kir6.2/SUR2A channels heterologously expressed in COSm6 cells. In mouse atrial myocytes, both spontaneously activated and diazoxide-activated K(ATP) currents were effectively inhibited by 10 μM HMR 1098. By contrast, in ventricular myocytes, pinacidil-activated K(ATP) currents were inhibited by HMR 1098 at a high concentration (100 μM) but not at a low concentration (10 μM). Consistent with this finding, HMR 1098 inhibits (86)Rb(+) effluxes through Kir6.2/SUR1 more effectively than Kir6.2/SUR2A channels in COSm6 cells. In excised inside-out patches, HMR 1098 inhibited Kir6.2/SUR1 channels more effectively, particularly in the presence of MgADP and MgATP (mimicking physiological stimulation). Finally, dose-dependent enhancement of insulin secretion from pancreatic islets and decrease of blood glucose level confirm that HMR 1098 is an inhibitor of Kir6.2/SUR1-composed K(ATP) channels.
Sigma-Aldrich
HMR-1098, ≥98% (HPLC)