Merck
  • Three-dimensional bacterial cellulose/polydopamine/TiO2 nanocomposite membrane with enhanced adsorption and photocatalytic degradation for dyes under ultraviolet-visible irradiation.
Three-dimensional bacterial cellulose/polydopamine/TiO2 nanocomposite membrane with enhanced adsorption and photocatalytic degradation for dyes under ultraviolet-visible irradiation.
Journal of colloid and interface science (2019-12-13)
Luyu Yang, Chuntao Chen, Ying Hu, Feng Wei, Jian Cui, Yuxiang Zhao, Xuran Xu, Xiao Chen, Dongping Sun
ABSTRACT
Cleaner production of photocatalyst with efficient property and stable reusability is of great importance for the elimination of organic pollutants in wastewater. Herein, we present a bacterial cellulose (BC)-based nanocomposite membrane with enhanced adsorption and photocatalytic degradation for dyes under UV radiation, by using BC nanofibers as a three-dimensional soft template, coated with the polydoamine (PDA) as a functional layer and a protective agent for immobilization of titanium dioxide (TiO2) nanoparticles. Compared with commercial P25, the as-prepared BC/PDA/TiO2 composite presented higher adsorption capacity for methyl orange, Rhodamine B and methylene blue, and the photocatalytic properties within 30 min after irradiation were further improved. BC/PDA/TiO2 also showed good stability, proved by the 5.5% reduction in photocatalytic capability after five cyclic tests. We expect our design could provide a facile and green approach with excellent photo-degradation performance for organic pollutants, providing further applications for photocatalysis and wastewater treatment.
Sigma-Aldrich
Ammonium hexafluorotitanate, 99.99% trace metals basis