Merck
  • Thymosin α1 represents a potential potent single-molecule-based therapy for cystic fibrosis.
Thymosin α1 represents a potential potent single-molecule-based therapy for cystic fibrosis.
Nature medicine (2017-04-11)
Luigina Romani, Vasilis Oikonomou, Silvia Moretti, Rossana G Iannitti, Maria Cristina D'Adamo, Valeria R Villella, Marilena Pariano, Luigi Sforna, Monica Borghi, Marina M Bellet, Francesca Fallarino, Maria Teresa Pallotta, Giuseppe Servillo, Eleonora Ferrari, Paolo Puccetti, Guido Kroemer, Mauro Pessia, Luigi Maiuri, Allan L Goldstein, Enrico Garaci
ABSTRACT
Cystic fibrosis (CF) is caused by mutations in the gene encoding the cystic fibrosis transmembrane conductance regulator (CFTR) that compromise its chloride channel activity. The most common mutation, p.Phe508del, results in the production of a misfolded CFTR protein, which has residual channel activity but is prematurely degraded. Because of the inherent complexity of the pathogenetic mechanisms involved in CF, which include impaired chloride permeability and persistent lung inflammation, a multidrug approach is required for efficacious CF therapy. To date, no individual drug with pleiotropic beneficial effects is available for CF. Here we report on the ability of thymosin alpha 1 (Tα1)-a naturally occurring polypeptide with an excellent safety profile in the clinic when used as an adjuvant or an immunotherapeutic agent-to rectify the multiple tissue defects in mice with CF as well as in cells from subjects with the p.Phe508del mutation. Tα1 displayed two combined properties that favorably opposed CF symptomatology: it reduced inflammation and increased CFTR maturation, stability and activity. By virtue of this two-pronged action, Tα1 has strong potential to be an efficacious single-molecule-based therapeutic agent for CF.
Sigma-Aldrich
Anti-Indoleamine 2,3-dioxygenase Antibody, clone 10.1, clone 10.1, Chemicon®, from mouse
Sigma-Aldrich
Thymosin α1 bovine, ≥90% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution