Merck
  • Common and unique strategies of male killing evolved in two distinct Drosophila symbionts.
Common and unique strategies of male killing evolved in two distinct Drosophila symbionts.
Proceedings. Biological sciences (2018-03-23)
Toshiyuki Harumoto, Takema Fukatsu, Bruno Lemaitre
ABSTRACT
Male killing is a selfish reproductive manipulation caused by symbiotic bacteria, where male offspring of infected hosts are selectively killed. The underlying mechanisms and the process of their evolution are of great interest not only in terms of fundamental biology, but also their potential applications. The two bacterial Drosophila symbionts, Wolbachia and Spiroplasma, have independently evolved male-killing ability. This raises the question whether the underlying mechanisms share some similarities or are specific to each bacterial species. Here, we analyse pathogenic phenotypes of D. bifasciata infected with its natural male-killing Wolbachia strain and compare them with those of D. melanogaster infected with male-killing Spiroplasma We show that male progeny infected with the Wolbachia strain die during embryogenesis with abnormal apoptosis. Interestingly, male-killing Wolbachia infection induces DNA damage and segregation defects in the dosage-compensated chromosome in male embryos, which are reminiscent of the phenotypes caused by male-killing Spiroplasma in D. melanogaster By contrast, host neural development seems to proceed normally unlike male-killing Spiroplasma infection. Our results demonstrate that the dosage-compensated chromosome is a common target of two distinct male killers, yet Spiroplasma uniquely evolved the ability to damage neural tissue of male embryos.
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys16) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Heat Shock Protein 60 antibody produced in mouse, clone LK2, ascites fluid