- Prevalence of Streptococcus pneumoniae serotype 6C among invasive and carriage isolates in metropolitan Salvador, Brazil, from 1996 to 2007.
Prevalence of Streptococcus pneumoniae serotype 6C among invasive and carriage isolates in metropolitan Salvador, Brazil, from 1996 to 2007.
Diagnostic microbiology and infectious disease (2009-08-28)
Leila C Campos, Maria da Gloria S Carvalho, Bernard W Beall, Soraia M Cordeiro, Daniele Takahashi, Mitermayer G Reis, Albert I Ko, Joice N Reis
PMID19709842
ABSTRACT
The newly described Streptococcus pneumoniae serotype 6C accounted for 2.3% (16/709) of meningitis cases and 3.2% (3/95) of nasopharyngeal isolates from healthy individuals in Brazil. The strains were multidrug resistant (18.8%) and genetically diverse. Despite low serotype 6C prevalence, continuous surveillance is necessary to guide vaccine strategies.