Merck
  • DABCO-bis(sulfur dioxide), DABSO, as a convenient source of sulfur dioxide for organic synthesis: utility in sulfonamide and sulfamide preparation.
DABCO-bis(sulfur dioxide), DABSO, as a convenient source of sulfur dioxide for organic synthesis: utility in sulfonamide and sulfamide preparation.
Organic letters (2011-08-27)
Holly Woolven, Carlos González-Rodríguez, Isabel Marco, Amber L Thompson, Michael C Willis
ABSTRACT
The charge-transfer complex generated from the combination of DABCO and sulfur dioxide, DABSO, is a bench-stable colorless solid suitable for use in organic synthesis as a replacement for gaseous sulfur dioxide. The complex can be combined with Grignard reagents to form sulfinates, which can then be converted in situ to a series of sulfonamides. Alternatively, reaction with anilines and iodine leads to the formation of a series of sulfamides. Cheletropic addition between DABSO and 2,3-dimethylbutadiene provides the corresponding sulfolene.
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct, ≥95% (sulfur, elemental analysis)
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Dabco® 33-LV