Merck
  • Antimicrobial activity and safety applications of meso-tetra(4-pyridyl)platinum(II) porphyrin.
Antimicrobial activity and safety applications of meso-tetra(4-pyridyl)platinum(II) porphyrin.
Microbial pathogenesis (2018-12-24)
Leonardo Quintana Soares Lopes, Andiara Prates Ramos, Priscila Marquezan Copetti, Thiago Vargas Acunha, Bernardo Almeida Iglesias, Roberto Christ Vianna Santos, Alencar Kolinski Machado, Michele Rorato Sagrillo
ABSTRACT
Tetra-platinated(II) porphyrin hexafluorophosphate compound (4-PtTPyPor) was synthetized and along 5,10,15,20-tetrakis(4-pyridyl)porphyrin (4-TPyPor), evaluated about the antimicrobial activity and safety. The effect was evaluated with and without light exposition. The antimicrobial activity was analyzed by microdilution and growth curve method. The assays showed an increase of antimicrobial potential caused by porphyrins with light exposition comparing the treatment without light irradiation. The biocompatibility was tested by MTT, ROS production, dsDNA on culture medium and hemolysis. All platinum porphyrin concentrations showed hemolytic activity under light exposition. The ROS measurement doesn't showed statistic difference between treatments and control. The picogreen assay demonstrates a reduction of dsDNA on culture medium with cells treated with porphyrins under light irradiation. The study demonstrated that the platinated porphyrins might be promising microbial photodynamic inactivation with potential applications in wastewater treatment, biofilm control and bioremediation.