- Intracerebroventricular administration of lupus serum induces microglia activation and leukocyte adhesion in the cerebromicrovasculature of mice.
Intracerebroventricular administration of lupus serum induces microglia activation and leukocyte adhesion in the cerebromicrovasculature of mice.
Central nervous system (CNS) involvement is commonly seen in the patients with system lupus erythematosus (SLE). Mechanisms underlying CNS damage in SLE remain largely unknown. Accumulating evidence suggest that activation of microglia in CNS plays an important role in the inflammatory responses in neurological diseases. The aim of this study is to examine the involvement of microglia in the CNS inflammatory responses induced by circulating serum of SLE patients. We performed intracerebroventricular (ICV) injection of serums collected from SLE patients or healthy controls to mice, and examined phenotypic changes of microglia, the levels of cytokines, chemokine and adhesion molecules in the brain. Intravital microscopy was used to observe leukocyte rolling and adhesion in the cerebromicrovasculature. We further examined whether minocycline can block inflammatory responses induced by SLE serum. In vitro experiments were conducted to examine whether IgGs from the sera of SLE patients or healthy control can activate the primary cultured microglia. We found that ICV injection of SLE serum increases morphological activation of microglia in the cortex and hippocampus. Inflammatory mediators including pro-inflammatory cytokines (IL-1, IL-6 and TNF-α), chemokine (CCL2 and CCL5) and adhesion molecules (P-selectin and ICAM-1) were significantly elevated in the brains of SLE-serum-treated mice. Using intravital microscopy, we demonstrated that SLE serum promotes leukocyte rolling and adhesion. Furthermore, suppression of microglia activation by systemically using minocycline could decrease the levels of inflammatory molecular, and prevent leukocyte rolling and adhesion. The in vitro experiments revealed that IgG from SLE sera could be engulfed by microglia and stimulated the microglia to secret pro-inflammatory cytokines. Our data suggest that the activation of microglia, which promotes leukocyte adhesion to the brain microvasculature, is an important pathological mechanism of CNS involvement in SLE.