Merck
  • The Novel p38 Inhibitor, Pamapimod, Inhibits Osteoclastogenesis and Counteracts Estrogen-Dependent Bone Loss in Mice.
The Novel p38 Inhibitor, Pamapimod, Inhibits Osteoclastogenesis and Counteracts Estrogen-Dependent Bone Loss in Mice.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2019-01-08)
Xiangde Zhao, Lei Ning, Ziang Xie, Zhiwei Jie, Xiang Li, Xinyu Wan, Xuewu Sun, Bao Huang, Pan Tang, Shuying Shen, An Qin, Yan Ma, Lu Song, Shunwu Fan, Shuanglin Wan
ABSTRACT
Pamapimod (PAM) is a novel selective p38 mitogen-activated protein (MAP) kinase inhibitor proved to be effective in rheumatoid arthritis in phase 2 clinical trial. However, its effect on osteoclast-associated osteoporosis and the underlying mechanisms remain unclear. In this study, we showed that PAM suppressed receptor activator of nuclear factor-κB ligand (RANKL)-induced osteoclast formation via inhibition of p38 phosphorylation and subsequent c-Fos and nuclear factor of activated T cells c1 (NFATc1) expression. In addition, the downregulated NFATc1 leads to reduced expression of its targeting gene disintegrin and metalloproteinase domain-containing protein 12 (ADAM12), which was further proven to be critical for osteoclastic bone resorption. Therefore, we treated ovariectomized (OVX) mice with PAM and revealed a protective effect of PAM on osteoporosis in vivo. In conclusion, our results demonstrated PAM can prevent OVX-induced bone loss through suppression of p38/NFATc1-induced osteoclast formation and NFATc1/ADAM12-associated bone resorption. © 2018 American Society for Bone and Mineral Research.