Merck
  • Synthesis of Cyanate Esters Based on Mono-O-Methylated Bisphenols with Sulfur-Containing Bridges.
Synthesis of Cyanate Esters Based on Mono-O-Methylated Bisphenols with Sulfur-Containing Bridges.
Molecules (Basel, Switzerland) (2019-01-10)
Andrey Galukhin, Roman Nosov
ABSTRACT
We described a synthetic approach to bisphenol-based monocyanate esters based on mono-O-methylation of parental bisphenols followed by cyanation of the residual phenolic hydroxyl. Structures of the synthesized compounds were determined by the application of IR, NMR ¹H and 13C spectroscopies, EI and MALDI mass spectrometry, and purity of the final product was controlled by HPLC. We showed that stability of the cyanate esters depends on the nature of the bridging group. Temperature range of thermally initiated cyclotrimerization of synthesized monocyanate ester, as well as reaction enthalpy, was determined by differential scanning calorimetry (DSC).