Merck
  • Radial profile detection of multiple spherical particles in contact with interacting surfaces.
Radial profile detection of multiple spherical particles in contact with interacting surfaces.
PloS one (2019-04-03)
Johannes Waschke, Tilo Pompe, David Rettke, Stephan Schmidt, Mario Hlawitschka
ABSTRACT
Adhesive interactions of soft materials play an important role in nature and technology. Interaction energies can be quantified by determining contact areas of deformable microparticles with the help of reflection interference contrast microscopy (RICM). For high throughput screening of adhesive interactions, a method to automatically evaluate large amounts of interacting microparticles was developed. An image is taken which contains circular interference patterns with visual characteristics that depend on the probe's shape due to its surface interaction. We propose to automatically detect radial profiles in images, and to measure the contact radius and size of the spherical probe, allowing the determination of particle-surface interaction energy in a simple and fast imaging and image analysis setup. To achieve this, we analyze the image gradient and we perform template matching that utilizes the physical foundations of reflection interference contrast microscopy.