Merck
  • The role of inosine-5'-monophosphate dehydrogenase in thiopurine metabolism in patients with inflammatory bowel disease.
The role of inosine-5'-monophosphate dehydrogenase in thiopurine metabolism in patients with inflammatory bowel disease.
Therapeutic drug monitoring (2011-02-12)
Sofie Haglund, Svante Vikingsson, Jan Söderman, Ulf Hindorf, Christer Grännö, Margareta Danelius, Sally Coulthard, Curt Peterson, Sven Almer
ABSTRACT
There is a large interindividual variability in thiopurine metabolism. High concentrations of methylthioinosine-5'-monophosphate (meTIMP) and low concentrations of 6-thioguanine nucleotides (6-TGNs) have been associated with a lower response rate and an increased risk of adverse events. In this study, the role of inosine-5'-monophosphate dehydrogenase (IMPDH) for differences in metabolite patterns of thiopurines was investigated. IMPDH activity and thiopurine metabolite concentrations were determined in patients with inflammatory bowel disease and a normal thiopurine methyltransferase (TPMT) phenotype and meTIMP/6-TGN concentration ratio > 20 (n = 26), in patients with a metabolite ratio ≤ 20 (n = 21), in a subgroup with a metabolite ratio <4 (n = 6), and in 10 patients with reduced TPMT activity. In vitro studies were conducted on human embryonic kidney cells (HEK293) with genetically engineered IMPDH and TPMT activities. Patients with metabolite ratios >20 had lower IMPDH activity than those with ratios ≤ 20 (P < 0.001). Metabolite ratios >20 were only observed in patients with normal TPMT activity. Downregulation of IMPDH activity in HEK293 cells was associated with an increase in the concentration of meTIMP (fold change: 17 up to 93, P < 0.001) but, unexpectedly, also of 6-thioguanosine monophosphate (fold change: 2.6 up to 5.0, P < 0.001). These data question the general view of IMPDH as the rate-limiting enzyme in the phosphorylation of thiopurines. Investigations of other mechanisms are needed to more fully explain the various metabolite patterns and outcomes in patients under treatment.
Sigma-Aldrich
Inosine 5′-monophosphate disodium salt hydrate, ≥99.0% (HPLC)
Sigma-Aldrich
6-Methylmercaptopurine riboside, ≥99% (HPLC)