Merck
  • Bivalency and epitope specificity of a high-affinity IgG3 monoclonal antibody to the Streptococcus group A carbohydrate antigen. Molecular modeling of a Fv fragment.
Bivalency and epitope specificity of a high-affinity IgG3 monoclonal antibody to the Streptococcus group A carbohydrate antigen. Molecular modeling of a Fv fragment.
Carbohydrate research (2000-03-21)
J B Pitner, W F Beyer, T M Venetta, C Nycz, M J Mitchell, S L Harris, J R Mariño-Albernas, F I Auzanneau, F Forooghian, B M Pinto
ABSTRACT
The binding of Strep 9, a mouse monoclonal antibody (mAb) of the IgG3 subclass directed against the cell-wall polysaccharide of Group A Streptococcus (GAS), has been characterized. The intact antibody and proteolytic fragments of Strep 9 bind differently to GAS: the intact mAb and F(ab)2' have greater affinity for the carbohydrate epitope than the monomeric Fab or F(ab)'. A mode of binding in which Strep 9 binds bivalently to portions of the polysaccharide on adjacent chains on GAS is proposed. A competitive ELISA protocol using a panel of carbohydrate inhibitors shows that the branched trisaccharide, beta-D-GlcpNAc-(1-->3)-[alpha-L-Rhap-(1-->2)]-alpha-L-Rhap, and an extended surface are key components of the epitope recognized by Strep 9. Microcalorimetry measurements with the mAb and two synthetic haptens, a tetrasaccharide and a hexasaccharide, show enthalpy-entropy compensation as seen in other oligosaccharide-protein interactions. Molecular modeling of the antibody variable region by homology modeling techniques indicates a groove-shaped combining site that can readily accommodate extended surfaces. Visual docking of an oligosaccharide corresponding to the cell-wall polysaccharide into the site provides a putative model for the complex, in which a heptasaccharide unit occupies the site and the GlcpNAc residues of two adjacent branched trisaccharide units occupy binding pockets within the groove-shaped binding site.
Sigma-Aldrich
N-Acetyl-D-glucosamine–Agarose, saline suspension