Merck
  • Study of the chemical derivatization of zearalenone and its metabolites for gas chromatography-mass spectrometry analysis of environmental samples.
Study of the chemical derivatization of zearalenone and its metabolites for gas chromatography-mass spectrometry analysis of environmental samples.
Journal of chromatography. A (2008-04-09)
Said Kinani, Stéphane Bouchonnet, Sophie Bourcier, Jean-Marc Porcher, Sélim Aït-Aïssa
ABSTRACT
This study compares different silylation procedures of zearalenone and its metabolites: alpha-zearalenol, beta-zearalenol, zearalanone, alpha-zearalanol and beta-zearalanol for gas chromatography-mass spectrometry (GC-MS) analysis. Four silylating agents among the most frequently used to derivatize polar organic compounds were tested: N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA), N-methyl-N-trimethylsilyltrifluoroacetamide (MSTFA), N,N-diethyltrimethylsilylamine (TMSDEA) and a commercial mixture of N,O-bis(trimethylsilyl)acetamide, trimethylchlorosilane and N-trimethylsilyimidazole. Previous studies showed that the addition of polar and/or basic solvents can significantly improve the yield of a reaction of derivatization. In this work, four solvents were tested: pyridine, dimethylformamide, acetonitrile and acetone. The influence of each solvent was studied as a function of the silylating agent/solvent ratio. The influences of the temperature and of the reaction time on the reaction yields were also evaluated. A GC-MS quantitation method associating methanol chemical ionization and selected ion storage with three ions was developed and successfully tested on a reconstituted sediment spiked in zearalenone and its metabolites.
Sigma-Aldrich
N,N-Diethyltrimethylsilylamine, 97%