Merck
  • The microtubule-associated protein tau cross-links to two distinct sites on each alpha and beta tubulin monomer via separate domains.
The microtubule-associated protein tau cross-links to two distinct sites on each alpha and beta tubulin monomer via separate domains.
Biochemistry (1999-01-28)
M F Chau, M J Radeke, C de Inés, I Barasoain, L A Kohlstaedt, S C Feinstein
ABSTRACT
The interaction between tubulin subunits and microtubule-associated proteins (MAPs) such as tau is fundamental for microtubule structure and function. Previous work has suggested that the "microtubule binding domain" of tau (composed of three or four imperfect 18-amino acid repeats, separated by 13- or 14-amino acid inter-repeat regions) can bind to the C-terminal ends of both alpha and beta tubulin monomers. Here, using covalent cross-linking strategies, we demonstrate that there are two distinct tau cross-linking sites (designated as "C-terminal" and "internal") on each alpha and beta tubulin monomer. The C-terminal tau cross-linking site is located within the 12 C-terminal amino acids of both alpha and beta tubulin, while the internal tau cross-linking site is located within the C-terminal one-third of alpha and beta tubulin but not within the last 12 amino acids. In addition, we show that tau cross-links to the C-terminal site via its repeat 1 and/or the R1-R2 inter-repeat. The cross-linking of tau to the internal site is mediated by some subset of its other repeat units. Integrating these and earlier data with the 3.7 A resolution model of the alphabeta tubulin dimer recently presented by E. Nogales et al. [(1998), Nature 391, 199-203], we propose a new model for the tau-microtubule interaction.
Sigma-Aldrich
Tubulin, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution