Merck
  • Non-invasive estimation of 10 B-4-borono-L-phenylalanine-derived boron concentration in tumors by PET using 4-borono-2-18 F-fluoro-phenylalanine.
Non-invasive estimation of 10 B-4-borono-L-phenylalanine-derived boron concentration in tumors by PET using 4-borono-2-18 F-fluoro-phenylalanine.
Cancer science (2018-03-03)
Mitsuyoshi Yoshimoto, Natsuki Honda, Hiroaki Kurihara, Kenta Hiroi, Satoshi Nakamura, Masashi Ito, Naoto Shikano, Jun Itami, Hirofumi Fujii
ABSTRACT
In boron neutron capture therapy (BNCT), 10 B-4-borono-L-phenylalanine (BPA) is commonly used as a 10 B carrier. PET using 4-borono-2-18 F-fluoro-phenylalanine (18 F-FBPA PET) has been performed to estimate boron concentration and predict the therapeutic effects of BNCT; however, the association between tumor uptake of 18 F-FBPA and boron concentration in tumors remains unclear. The present study investigated the transport mechanism of 18 F-FBPA and BPA, and evaluated the utility of 18 F-FBPA PET in predicting boron concentration in tumors. The transporter assay revealed that 2-aminobicyclo-(2.2.1)-heptane-2-carboxylic acid, an inhibitor of the L-type amino acid transporter, significantly inhibited 18 F-FBPA and 14 C-4-borono-L-phenylalanine (14 C-BPA) uptake in FaDu and LN-229 human cancer cells. 18 F-FBPA uptake strongly correlated with 14 C-BPA uptake in 7 human tumor cell lines (r = .93; P < .01). PET experiments demonstrated that tumor uptake of 18 F-FBPA was independent of the administration method, and uptake of 18 F-FBPA by bolus injection correlated well with BPA uptake by continuous intravenous infusion. The results of this study revealed that evaluating tumor uptake of 18 F-FBPA by PET was useful for estimating 10 B concentration in tumors.
Sigma-Aldrich
α-(Methylamino)isobutyric acid, ≥97% (titration)