
Vindesine sulfate

Synonym(s):
Vindesine sulfate salt, Desacetylvinblastine amide sulfate salt
Linear Formula:
C43H55N5O7 · H2SO4
CAS Number:
Molecular Weight:
852.00
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing