
Pifithrin-μ

Synonym(s):
2-Phenylethynesulfonamide, PFTμ, Pifithrin-μ
Empirical Formula (Hill Notation):
C8H7NO2S
CAS Number:
Molecular Weight:
181.21
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2