Skip to Content
Merck
Octaethylene glycol monohexadecyl ether BioXtra, ≥98.0% (GC)

Octaethylene glycol monohexadecyl ether

Octaethylene glycol monohexadecyl ether BioXtra, ≥98.0% (GC)
Synonym(s):
C16E8, Hexadecyl octaethylene glycol ether, Hexadecyloctaglycol
Linear Formula:
CH3(CH2)15(OCH2CH2)8OH
CAS Number:
Molecular Weight:
594.86

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing