Skip to Content
Merck
Cyanocobalamin British Pharmacopoeia (BP) Assay Standard

Cyanocobalamin

Cyanocobalamin British Pharmacopoeia (BP) Assay Standard
Synonym(s):
Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin
Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique