Crospovidone
Synonym(s):
Polyvinylpyrrolidone, PVP, Polyvidone, Povidone
Linear Formula:
(C6H9NO)n
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing