Skip to Content
Merck
cis-3-Hexen-1-ol analytical standard

cis-3-Hexen-1-ol

cis-3-Hexen-1-ol analytical standard
Linear Formula:
C2H5CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing