![6-O-Methylguanine 97%](/deepweb/assets/sigmaaldrich/product/structures/248/885/89148b73-faa1-4a1b-a528-1eb4b45703b0/640/89148b73-faa1-4a1b-a528-1eb4b45703b0.png)
6-O-Methylguanine
![6-O-Methylguanine 97%](/deepweb/assets/sigmaaldrich/product/structures/248/885/89148b73-faa1-4a1b-a528-1eb4b45703b0/640/89148b73-faa1-4a1b-a528-1eb4b45703b0.png)
Synonym(s):
2-Amino-6-methoxypurine, 6-Methoxyguanine
Empirical Formula (Hill Notation):
C6H7N5O
CAS Number:
Molecular Weight:
165.15
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing