Skip to Content
Merck
vibrant-m

4,4′-(2-Pyridinylmethylene)bisphenol

vibrant-m
Synonym(s):
4,4′-(Pyridin-2-ylmethylene)diphenol, DDPM, Deacetylbisacodyl
Empirical Formula (Hill Notation):
C18H15NO2
CAS Number:
Molecular Weight:
277.32

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1