![[3-(4-methoxy-phenyl)-isoxazol-5-yl]-methanol AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/119/604/3c9c972a-d615-421f-9c6a-e3a432c30b5b/640/3c9c972a-d615-421f-9c6a-e3a432c30b5b.png)
[3-(4-methoxy-phenyl)-isoxazol-5-yl]-methanol
![[3-(4-methoxy-phenyl)-isoxazol-5-yl]-methanol AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/119/604/3c9c972a-d615-421f-9c6a-e3a432c30b5b/640/3c9c972a-d615-421f-9c6a-e3a432c30b5b.png)
Empirical Formula (Hill Notation):
C11H11NO3
Molecular Weight:
205.21
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing