Merck
  • Purification of Two Novel Sugar Acid-binding Lectins from Haplomitrium Mnioides (bryophyte, Plantae) and their Preliminary Characterization.
Purification of Two Novel Sugar Acid-binding Lectins from Haplomitrium Mnioides (bryophyte, Plantae) and their Preliminary Characterization.
Applied biochemistry and biotechnology (2016-08-11)
Hiroaki Masuzaki, Masahiro Hosono, Kazuo Nitta
ABSTRACT
Two novel sugar acid-binding lectins were purified from Haplomitrium mnioides (Lindb.) Schust. using a procedure consisting of ammonium sulfate precipitation, G-50 gel filtration, hydroxyapatite chromatography, and HW-50 gel filtration. We reported their partial physicochemical properties: molecular weight, affinity for carbohydrates and organic acids, pH stability, and dependence of their hemagglutination activity on metal ions. We also determined their N-terminal amino acid sequences. H. mnioides lectins (HMLs) were monomers (one with a molecular weight of approximately 27 kDa, and the other with a molecular weight of approximately 105 kDa) under both nonreducing and reducing conditions. They were named HML27 and HML105, respectively. Both HMLs had an affinity for N-acetylneuraminic acid, D-glucuronic acid, D-glucaric acid, bovine submaxillary mucin, heparin, and organic acids, such as citrate, 2-oxoglutaric acid, and D-2-hydroxyglutarate. Furthermore, HML27 had an affinity for α-D-galacturonic acid, D-malate, L-malate, and pyruvate, while HML105 had an affinity for D-gluconic acid. HML27 and HML105 are novel plant lectins: they have an affinity for sugar acids and organic acids and specifically recognize the carboxyl group, and there is no homology between their N-terminal amino acid sequences and those of the previously described lectins and agglutinins.
Sigma-Aldrich
D-(+)-Fucose, ≥97% (GC)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG