Merck
  • 1,3,5-Trisubstituted benzenes as fluorescent photoaffinity probes for human carbonic anhydrase II capture.
1,3,5-Trisubstituted benzenes as fluorescent photoaffinity probes for human carbonic anhydrase II capture.
Chemical communications (Cambridge, England) (2013-02-01)
Partha Sarathi Addy, Baisakhee Saha, N D Pradeep Singh, Amit K Das, Jacob T Bush, Clarisse Lejeune, Christopher J Schofield, Amit Basak
ABSTRACT
The synthesis of small molecule based 1,3,5-trisubstituted benzenes for photo-mediated capture of human carbonic anhydrase II with visualisation by fluorescence is described.
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, analytical standard