Merck
  • Domperidone: limited benefits with significant risk for sudden cardiac death.
Domperidone: limited benefits with significant risk for sudden cardiac death.
Journal of cardiovascular pharmacology (2012-11-29)
Luc M Hondeghem
ABSTRACT
Domperidone (antinausea/vomiting agent) was recently shown by several groups to increase sudden cardiac death (SCD). Drug-induced disturbances of cardiac repolarization may be a major mechanism. Experiments were executed in isolated female rabbit hearts perfused for 150 minutes with domperidone 30, 60, or 100 nM. Domperidone significantly prolonged the action potential duration: +9% at 30 nM, +32% at 60 nM, and +48% at 100 nM. Domperidone induced significant disturbances of repolarization in 83% of hearts at 60 nM and in 100% at 100 nM, including early afterdepolarizations and polymorphic ventricular tachycardia. Maximum therapeutic free drug plasma concentration of domperidone (19 nM) yields a safety index of only ∼2.5, that is, 12-fold below the accepted minimum. Gastrointestinal benefits and risks for SCD were derived from the literature. The defined daily dose of domperidone (30 mg/day) fails to show unequivocal gastrointestinal benefits beyond a placebo effect. In contrast, 5 of 5 population-based studies show that oral domperidone significantly increases the odds ratio for SCD to 2.8 (1.53-6.21) and it increases sharply above 30 mg/day. Because domperidone has placebo-like benefits but is associated with increased SCD and a narrow safety margin, it should not be used in medicine.
Product Description
Domperidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Domperidone, powder, ≥98% (HPLC)