Merck
  • Generation of monoclonal antibody fragments binding the native γ-secretase complex for use in structural studies.
Generation of monoclonal antibody fragments binding the native γ-secretase complex for use in structural studies.
Biochemistry (2012-10-17)
Jean-René Alattia, Claude Schweizer, Matthias Cacquevel, Mitko Dimitrov, Lorène Aeschbach, Mustapha Oulad-Abdelghani, Patrick C Fraering
ABSTRACT
A detailed understanding of γ-secretase structure is crucially needed to elucidate its unique properties of intramembrane protein cleavage and to design therapeutic compounds for the safe treatment of Alzheimer's disease. γ-Secretase is an enzyme complex composed of four membrane proteins, and the scarcity of its supply associated with the challenges of crystallizing membrane proteins is a major hurdle for the determination of its high-resolution structure. This study addresses some of these issues, first by adapting CHO cells overexpressing γ-secretase to growth in suspension, thus yielding multiliter cultures and milligram quantities of highly purified, active γ-secretase. Next, the amounts of γ-secretase were sufficient for immunization of mice and allowed generation of Nicastrin- and Aph-1-specific monoclonal antibodies, from which Fab fragments were proteolytically prepared and subsequently purified. The amounts of γ-secretase produced are compatible with robot-assisted crystallogenesis using nanoliter technologies. In addition, our Fab fragments bind exposed regions of native γ-secretase in a dose-dependent manner without interfering with its catalytic properties and can therefore be used as specific tools to facilitate crystal formation.
Sigma-Aldrich
CHAPSO, ≥98%
Sigma-Aldrich
CHAPSO, BioXtra