Merck
Hypolipidemic activity of Indan--1--acetic acids.
Nepal Medical College journal : NMCJ (2006-07-11)
Manoranjan Adak, Joyant Kumar Gupta
ABSTRACT
To study the hyoplipidemic activity of non-methoxy and methoxy substituted--alkyl indan acetic acids in normogenic and hypergenic animal models. The hyoplipidemic activity was evaluated using normogenic and hypergenic rat models. Indan acids synthesized in our laboratory (50 mg/kg) and the standard drug clofibrate (50 mg/kg) were used for this study. Dimethoxy substituted -methyl and -ethyl indan acetic acids (9 & 10) exhibited significant hypocholesterolemic activity in both animal models. Clofibrate showed cholesterol lowering activity of 18% in normogenic rats while test agents 9 and 10 produced a similar activity of 20% and 17% respectively. Liver cholesterol and liver weight of the animals tested were found normal in case of the test compounds while liver weight was increased by 15% in clofibrate treated rats. LDL and VLDL cholesterol levels were also affected by compounds No. 9 and 10. Compounds having a smaller alkyl groups (R = CH3, C2H5) at--carbon atom of the acetic acid moiety exhibited significant hypocholesterolemic activity.
Product Description
Supelco
Indan, analytical standard
Sigma-Aldrich
Indan, 95%