Merck
  • Efficacy of permethrin- and cyphenothrin-impregnated nettings against Culex pipiens pallens.
Efficacy of permethrin- and cyphenothrin-impregnated nettings against Culex pipiens pallens.
Journal of the American Mosquito Control Association (1992-03-01)
T Itoh, T Kurihara
ABSTRACT
Laboratory tests were conducted to determine the effect of different bed net materials, impregnated with permethrin or cyphenothrin, on Culex mosquitoes. Polyethylene, polyester and vinylon nettings were more efficacious than cotton or nylon.
Product Description
Supelco
Cyphenothrin, PESTANAL®, analytical standard, sum of isomers