Merck
  • Proteinase-activated receptor-2-induced colonic inflammation in mice: possible involvement of afferent neurons, nitric oxide, and paracellular permeability.
Proteinase-activated receptor-2-induced colonic inflammation in mice: possible involvement of afferent neurons, nitric oxide, and paracellular permeability.
Journal of immunology (Baltimore, Md. : 1950) (2003-04-19)
Nicolas Cenac, Rafael Garcia-Villar, Laurent Ferrier, Muriel Larauche, Nathalie Vergnolle, Nigel W Bunnett, Anne-Marie Coelho, Jean Fioramonti, Lionel Bueno
ABSTRACT
Activation of colonic proteinase-activated receptor-2 (PAR-2) provokes colonic inflammation and increases mucosal permeability in mice. The mechanism of inflammation is under debate and could be neurogenic and/or the consequence of tight-junction opening with passage of exogenous pathogens into the lamina propria. The present study aimed to further characterize the inflammatory effect of PAR-2 activation by investigating: 1) the role of NO, 2) the role of afferent neurons, and 3) a possible cause and effect relationship between colonic paracellular permeability changes and mucosal inflammation. Thus, intracolonic infusion to mice of the PAR-2-activating peptide, SLIGRL, increased both myeloperoxidase (MPO) activity and damage scores indicating colonic inflammation, and enhanced colonic permeability to (51)Cr-EDTA from 2 to 4 h after its infusion. NO synthase inhibitors, L-NAME and aminoguanidine, as well as the neurotoxin capsaicin and NK1, calcitonin gene-related peptide (CGRP) receptor antagonists, SR140333 and CGRP(8-37), prevented SLIGRL-induced MPO and damage score increases and permeability. In contrast, although the tight-junction blocker, 2,4,6-triaminopyrimidine, and the myosin L chain kinase inhibitor, ML-7, prevented SLIGRL-induced increase in permeability, they did not prevent MPO and damage score increases. Taken together our data show that both NO and capsaicin-sensitive afferent neurons are involved in PAR-2-mediated colonic inflammation and paracellular permeability increase. Nevertheless, the inflammation process is not a consequence of increased permeability which results at least in part from the activation of myosin L chain kinase.
Sigma-Aldrich
2,4,6-Triaminopyrimidine, 97%