Merck
  • The Role of Calcium-Sensing Receptors in Endothelin-1-Dependent Effects on Adult Rat Ventricular Cardiomyocytes: Possible Contribution to Adaptive Myocardial Hypertrophy.
The Role of Calcium-Sensing Receptors in Endothelin-1-Dependent Effects on Adult Rat Ventricular Cardiomyocytes: Possible Contribution to Adaptive Myocardial Hypertrophy.
Journal of cellular physiology (2016-09-24)
Elena Dyukova, Rolf Schreckenberg, Christoph Arens, Guzel Sitdikova, Klaus-Dieter Schlüter
ABSTRACT
Nitric oxide (NO)-deficiency as it occurs during endothelial dysfunction activates the endothelin-1 (ET-1) system and increases the expression of receptor activity modifying protein (RAMP)-1 that acts as a chaperon for calcium-sensing receptors (CaR) that have recently been identified to improve cardiac function. Here, we hypothesized that ET-1 increases the cardiac expression of CaR and thereby induces an adaptive type of hypertrophy. Expressions of RAMP-1, endothelin receptors, and CaR were analyzed by RT-PCR in left ventricular tissues of L-NAME-treated rats. Effects of ET-1 on CaR expression and cell function (load free cell shortening) were analyzed in adult rat ventricular cardiomyocytes. siRNA directed against CaR and RAMP-1 was used to investigate a causal relationship. PD142893 and BQ788 were used to dissect the contribution of ETB1 , ETB2 , and ETA receptors. Non-specific NO synthase inhibition with L-Nitro arginine methyl ester (L-NAME) caused a cardiac upregulation of ETB receptors and CaR suggesting a paracrine effect of ET-1 on cardiomyocytes. Indeed, ET-1 induced the expression of CaR in cultured cardiomyocytes. Under these conditions, cardiomyocytes increased cell size (hypertrophy) but maintained normal function. Inhibition of ETA and ETB1 receptors led to ET-1-dependent reduction in cell shortening and attenuated up-regulation of CaR. Down-regulation of RAMP-1 reduced CaR responsiveness. In conclusion, ET-1 causes an adaptive type of hypertrophy by up-regulation of CaR in cardiomyocytes via ETA and/or ETB1 receptors. J. Cell. Physiol. 232: 2508-2518, 2017. © 2016 Wiley Periodicals, Inc.
Sigma-Aldrich
Anti-Calcium Sensing Receptor antibody produced in rabbit, affinity isolated antibody