Merck
Insulin-secreting INS-1E cells express functional TRPV1 channels.
Islets (2012-01-26)
Amanda Jabin Fågelskiöld, Kristina Kannisto, Anna Boström, Banina Hadrovic, Cecilia Farre, Mohamed Eweida, Kenneth Wester, Md Shahidul Islam
ABSTRACT
We have studied whether functional TRPV1 channels exist in the INS-1E cells, a cell type used as a model for β-cells, and in primary β-cells from rat and human. The effects of the TRPV1 agonists capsaicin and AM404 on the intracellular free Ca (2+) concentration ([Ca (2+)]i) in the INS-1E cells were studied by fura-2 based microfluorometry. Capsaicin increased [Ca (2+)]i in a concentration-dependent manner, and the [Ca (2+)]i increase was dependent on extracellular Ca (2+). AM404 also increased [Ca (2+)]i in the INS-1E cells. Capsazepine, a specific antagonist of TRPV1, completely blocked the capsaicin- and AM404-induced [Ca (2+)]i increases. Capsaicin did not increase [Ca (2+)]i in the primary β-cells from rat and human. Whole cell patch clamp configuration was used to record currents across the plasma membrane in the INS-1E cells. Capsaicin elicited inward currents that were inhibited by capsazepine. Western blot analysis detected TRPV1 proteins in the INS-1E cells and the human islets. Immunohistochemistry was used to study the expression of TRPV1, but no TRPV1 protein immunoreactivity was detected in the human islet cells and the human insulinoma cells. We conclude that the INS-1E cells, but not the primary β-cells, express functional TRPV1 channels.
Sigma-Aldrich
Anti-Vanilloid Receptor-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution