Merck
  • 2,2,6,6-Tetramethylpiperidin-1-yloxycarbonyl: A Protecting Group for Primary, Secondary, and Heterocyclic Amines.
2,2,6,6-Tetramethylpiperidin-1-yloxycarbonyl: A Protecting Group for Primary, Secondary, and Heterocyclic Amines.
Organic letters (2018-10-24)
Joseph R Lizza, Maximilian Bremerich, Stephanie R McCabe, Peter Wipf
ABSTRACT
The 2,2,6,6-tetramethylpiperidin-1-yloxycarbonyl (Tempoc) protecting group is readily introduced by the reaction of amines with a new acyl transfer reagent, 4-nitrophenyl (2,2,6,6-tetramethylpiperidin-1-yl) carbonate (NPTC). Tempoc has a reactivity profile that complements the commonly used t-butyloxycarbonyl (Boc) and benzyloxycarbonyl (Cbz) protecting groups. Deprotection can be achieved under mild reductive conditions with in situ generated Cu(I) species or by thermolytic cleavage at 135 °C. Mechanistic studies on the deprotection of Tempoc-indole suggest a combination of ionic and radical fragmentation pathways under thermal conditions.
Sigma-Aldrich
4-Nitrophenyl chloroformate, 96%
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
Copper(II) chloride, powder, 99%